Tlačiť
Návleky na boty, Secutex® per, PP/PE
Dodávateľ: ZVG BADEN WUERTEMBERG
30252-00 30251-00
113-0780EA 1084.24 EUR
113-0780 113-0779
Návleky na boty, Secutex® per, PP/PE
Jednorázové oděvy
These boot covers provide protection against fine dusts and splashes of aggressive liquid chemicals. Partial resistance to splashing.
  • Smooth, lint-free and abrasion-resistant material (PP spunbond with PE laminate)
  • Permeable to air and water vapour
  • Impervious to particles, with partial resistance to splashing and also anti-static
  • Choice of boot covers either elasticated or with ties
Ideal for use in the remediation of contaminated sites, building reconstruction or in asbestos abatement as well as in spray work in agriculture or in industry and car paint shops.
*One size fits all.
Certifikácie: Cat. III, type 5 + 6