Tlačiť
Defined foetal bovine serum, US origin, HyClone™
Dodávateľ: HyClone products (Cytiva)
SH30070.02 SH30070.03 SH30070.01 SH30070.03HI SH30070.02HI SH30070.01HI
HYCLSH30070.02EA 305.66 EUR
HYCLSH30070.02 HYCLSH30070.03 HYCLSH30070.01 HYCLSH30070.03HI HYCLSH30070.02HI SH30070.01HI
Defined foetal bovine serum, US origin, HyClone™
Séra
Defined foetal bovine serum (FBS) is the highest quality of FBS available and is very widely used by those cell culturist who have a concern for viral contaminants and require an extensive biochemical profile. Defined FBS is filtered through multiple 40 nm (0,04 µm) pore-size related filters, which are the most retentive filters used in commercial FBS production. Over 50 components are analysed and the results are included on the biochemical assay.
  • Filtered through serial 40nm pore-size rated filters
  • Maximize cell culture growth
  • U.S. sourced
Maximize cell culture growth with U.S.-sourced Cytiva HyClone Defined Fetal Bovine Serum (FBS), filtered through serial 40nm pore-size rated filters and tested for endotoxin, hemoglobin, and extensive biochemical profile assays.