Tlačiť
Screw caps, DURAN® PURE Premium STERICLIN®-Packed, DURAN®
Dodávateľ: DWK LIFE SCIENCES
892992806 892991304
215-2990EA 455.44 EUR
215-2990 215-3741
Screw caps, DURAN® PURE Premium STERICLIN®-Packed, DURAN®
Uzávěry Uzávěry k láhvím
STERICLIN®-packaging offers a sterile barrier system (SBS).
  • No pre-cleaning of the Premium cap is necessary
  • Ideal for dry heat sterilisation / depyrogenation
  • Can be sterilised directly in the STERICLIN® pouches
  • Sterile storage for up to 6 months
  • Pouch contains caps from a single manufacturing lot
These paper/film see-through pouches are compliant in accordance with DIN ISO 11607. The sterilisation-proof paper is an effective barrier against germs and bacteria, yet it permits the transmission of air, steam, or sterilisation gases. It‘s purpose is to preserve sterility after sterilisation. The three indicators for STEAM-, EO-, and FORM-sterilisation are applied on the side seam of the STERICLIN®-packaging . The SBS is widely used by medical and pharmaceutical companies and is the most used packing system in these fields. Premium caps are produced and assembled with previously specially cleaned PTFE-coated silicone seals and sealed in the SBS under Class 7 cleanroom conditions, in accordance with EN ISO 14644-1.