Tlačiť
Beakers, low form, Corning®
Dodávateľ: Corning
1000P-100 1000P-150 1000P-1L 1000P-250 1000P-2L 1000P-400 1000P-50 1000P-5L 1000P-600 1000P-25 1000P-3L
CORN1000P-100EA 38.99 EUR
CORN1000P-100 CORN1000P-150 CORN1000P-1L CORN1000P-250 CORN1000P-2L CORN1000P-400 CORN1000P-50 CORN1000P-5L CORN1000P-600 CORN1000P-25 CORN1000P-3L
Beakers, low form, Corning®
Kádinky
PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Autoclavable
With printed blue graduations.