Tlačiť
Polycarbonate erlenmeyer flasks with screw caps, Corning®
Dodávateľ: Corning
430183 430421 430422 431143 431144 431145 431146 431147 431252 431253 431255 431256 431401 431402 431403 431404 431405 431406 431407 431408 431684 431685
734-1820EA 605.7 EUR
734-1820 734-1832 734-1833 734-1885 734-1886 734-1887 734-1888 734-1889 734-1902 734-1903 734-1904 734-1905 734-4197 734-4198 734-4199 734-4200 734-4201 734-4202 734-4203 734-4204 734-2758 734-2759
Polycarbonate erlenmeyer flasks with screw caps, Corning®
Baňky Baňky dle Erlenmeyera
PC, optically clear.
  • Baffled or plain bottom options in all sizes (125 ml to 3 L)
  • Moulded-in graduations for accuracy
  • Vent cap option for continuous gas exchange while ensuring sterility and preventing leakage
  • Individually packaged and radiation sterilised for ease of use
  • Certified non-pyrogenic
Corning® baffled and plain Erlenmeyer flasks are ideal for shaker culture applications and storage.
Pozor: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.