Tlačiť
Replacement Erlenmeyer flask caps, Corning®
Dodávateľ: Corning
431339 431340 431363 431364 431372
734-1215EA 181.71 EUR
734-1215 734-1216 734-4194 734-4195 392-0292
Replacement Erlenmeyer flask caps, Corning®
Uzávěry Uzávěry k láhvím
Made of PP.
Replacement caps for Corning® 500 ml, 2 and 3 litre Erlenmeyer flasks are available separately. They are supplied sterile and individually packaged.
Pozor: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.