Tlačiť
Multiple well cell culture plates, Costar®
Dodávateľ: Corning
3471 3473 3506 3512 3513 3516 3526 3548 3527 3524 3335 3336 3337 3338 3736 3737 3738
734-1582EA 700.76 EUR
734-1582 734-1584 734-1596 734-1597 734-1598 734-1599 734-1605 734-1607 734-1606 734-1604 734-1210 734-1211 734-1212 734-4067 392-0213 392-0214 392-0215
Multiple well cell culture plates, Costar®
Mikrodestičky Destičky pro bunečné kultury
Clear PS, flat bottom, with lid. Available with a choice of surface treatments.
  • Lids with condensation rings to reduce contamination
  • Uniform footprint for ease of stacking
  • Individual alphanumeric codes for well identification
  • Sterilised by gamma radiation
  • Certified non-pyrogenic