Tlačiť
Cell culture dishes, untreated, Corning®
Dodávateľ: Corning
430588 430589 430591 430597
734-1707EA 263.94 EUR
734-1707 734-1708 734-1709 734-1710
Cell culture dishes, untreated, Corning®
Misky kultivační Misky pro tkáňové kultury
Optically clear PS. For applications where cell attachment is not desired.
  • With stacking beads to aid handling
  • Supplied with vents to provide consistent gas exchange
  • Sterilised by gamma radiation
  • Certified non-pyrogenic