Tlačiť
Clear bottom black and white polystyrene microplates, 96 well, Corning®
Dodávateľ: Corning
3601 3603 3604 3610 3614 3615 3631 3632 3651 3903 3904 3884 3883 3882 3881 3880 3886 3885 3340 3843 3904BC
734-1608EA 1105.5 EUR
734-1608 734-1609 732-5516 734-1610 732-5518 732-5519 734-1612 734-1613 732-5523 734-1660 734-1661 734-4158 734-4157 734-4156 734-4155 734-4154 734-4160 734-4159 734-4069 734-2293 CORN3904BC
Clear bottom black and white polystyrene microplates, 96 well, Corning®
Mikrodestičky Mikrodestičky a stripy pro imunologii
PS, flat bottom.
  • Bottoms are 60% thinner than conventional PS plates, resulting in lower background fluorescence and enabling readings down to 340 nm
  • Opaque walls prevent well-to-well crosstalk
  • Optically clear flat bottom profile permits direct microscope viewing
NBS™ = Non Binding Surface.