Tlačiť
384-well clear polystyrene microplates, Corning®
Dodávateľ: Corning
3700 3701 3702 3680 3640 3702BC 3844
732-5537EA 894.91 EUR
732-5537 732-5538 732-5539 732-5536 732-5520 734-4121 734-2294
384-well clear polystyrene microplates, Corning®
Mikrodestičky Destičky pro bunečné kultury
PS, flat bottom.
  • Total well volume of 112 μl; working well volume of 20 to 80 μl
  • Cell culture plates sterilised by gamma radiation and certified non pyrogenic
  • Universal optics NBS™ plate manufactured using an advanced polymer with high clarity and improved chemical resistance
  • Lids supplied with sterile plates, other lids can be ordered separately
White walled plates enhance luminescent signals and have low background luminescence and fluorescence. Black walled plates have low background fluorescence, minimal light scatter.