Tlačiť
General purpose gloves, ActivArmr® 80-409
Dodávateľ: Ansell
80-409/9 80-409/10 80-409/8
ANSE80-409/9EA 152.03 EUR
ANSE80-409/9 ANSE80-409/10 ANSE80-409/8
General purpose gloves, ActivArmr® 80-409
Rukavice
These seamless knitted nylon gloves with thermal yarns feature a full nitrile coating. They provide protection for medium mechanical applications as well as limited protection against cold.
  • Fully dipped
  • Fine and stretchable stitches for excellent comfort
  • Elastic knitwrist ensures snug fit
  • Excellent abrasion resistance
  • Covered in a layer resistant to polluted pieces being handled
Ideal for outdoor construction work, perishables warehousing, public maintenance, refrigeration and cooling plants, refuse collection and for shipping and receiving.
Certifikácie: EN 388:2016 (4121C), EN 511 (020); Cat. II
Food contact approved.