Tlačiť
Labels for BBP3x printers, type B-438, Defender™
Dodávateľ: BRADY
118076 118078 118077 118014
BRDY118076EA 234.26 EUR
BRDY118076 BRDY118078 BRDY118077 BRDY118014
Labels for BBP3x printers, type B-438, Defender™
Štítky
These tamper-evident metallic polyester labels have tamper-evident acrylic adhesive for smooth surfaces, withstand solvents and variable temperatures.
  • Leaves a CHECKERBOARD footprint when label is removed to prevent reuse
  • Ideal for product rating plates, product warranty labels and asset identification labels