Tlačiť
Anti-ICAM3 Mouse Monoclonal Antibody (CF640R) [clone: ICAM3/1019]
Dodávateľ: BIOTIUM
BNC401019-500 BNC401019-100
BNC401019-500EA 639.39 EUR
BNC401019-500 BNC401019-100
Anti-ICAM3 Mouse Monoclonal Antibody (CF640R) [clone: ICAM3/1019]
Protilátky
Recognizes an N-glycosylated glycoprotein of 120 kDa with intra-chain disulfide bonds, identified as CD50 or ICAM-3. CD50 is the major ligand for LFA-1 (CD11a/CD18) and may have signaling role to increase adhesion. It is expressed on thymocytes and T lymphocytes and is resistant to treatment with phosphatidylinositol (PI) phospholipase C. This MAb is excellent for staining of formalin/paraffin tissues.
Type: Primary
Antigen: ICAM3
Clonality: Monoclonal
Clone: ICAM3/1019
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human