Tlačiť
Anti-CD22 Mouse Monoclonal Antibody (CF488A) [clone: Myg13]
Dodávateľ: BIOTIUM
BNC880974-500 BNC880974-100
BNC880974-500EA 639.39 EUR
BNC880974-500 BNC880974-100
Anti-CD22 Mouse Monoclonal Antibody (CF488A) [clone: Myg13]
Protilátky
Recognizes a protein of 130-140 kDa, identified as CD22 (also known as BL-CAM). CD22 expression is restricted to normal and neoplastic B cells and is absent from other haemopoietic cell types. In B-cell ontogeny, CD22 is first expressed in the cytoplasm of pro-B and pre-B cells, and on the surface as B cells mature to become IgD . It is not expressed by plasma cells, CD22 is found highly expressed in follicular mantle and marginal zone B-cells, and while germinal center B-cells are relatively weak. CD22 is a member of the immunoglobulin superfamily and serves as an adhesion receptor for sialic acid-bearing ligands expressed on erythrocytes and all leukocyte classes. It also associates with tyrosine kinases and play a role in signal transduction and B-cell activation.
Type: Primary
Antigen: CD22
Clonality: Monoclonal
Clone: Myg13
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat