Tlačiť

Anti-COL3 Rabbit Polyclonal Antibody

Dodávateľ: Novus Biologicals
NOVUNB600-594EA 893.57 EUR
NOVUNB600-594
Anti-COL3 Rabbit Polyclonal Antibody
Protilátky
Rabbit Polyclonal Collagen III Antibody. Tested Applications: Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation. Tested Reactivity: Human, Mouse, Rat, Mammal.

Type: Primary
Antigen: COL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR