Tlačiť

Anti-IAPP Rabbit Polyclonal Antibody

Dodávateľ: Novus Biologicals
NOVUNBP1-06579EA 695.39 EUR
NOVUNBP1-06579
Anti-IAPP Rabbit Polyclonal Antibody
Protilátky
Rabbit Polyclonal Amylin Antibody. Tested Applications: Western Blot, ELISA, Immunohistochemistry. Tested Reactivity: Human, Mouse, Rat.

Type: Primary
Antigen: IAPP
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR