Tlačiť

Anti-CD28 Mouse Monoclonal Antibody (CF568) [clone: CB28]

Dodávateľ: BIOTIUM
BNC680384-500 BNC680384-100
BNC680384-500EA 639.39 EUR
BNC680384-500 BNC680384-100
Anti-CD28 Mouse Monoclonal Antibody (CF568) [clone: CB28]
Protilátky
Recognizes a glycoprotein of 44-88 kDa, which is identified as CD28. It is the critical T-cell co-stimulatory receptor which provides to the cell the important second activation signal by binding CD80 and CD86 that are expressed by antigen presenting cells. Besides its co-stimulation role, CD28 functions in preventing T-cells from anergic hyporesponsive state or from undergoing premature apoptotic cell death. CD28 is also expressed on human fetal NK cells and some NK cell lines, whereas on murine NK cells the CD28 expression is much broader.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: CB28
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR