Tlačiť
Anti-ITGAX Mouse Monoclonal Antibody (CF488A) [clone: HC1/1]
Dodávateľ: BIOTIUM
BNC880152-500 BNC880152-100
BNC880152-500EA 639.39 EUR
BNC880152-500 BNC880152-100
Anti-ITGAX Mouse Monoclonal Antibody (CF488A) [clone: HC1/1]
Protilátky
CD11c, also known as Integrin, alpha X (complement component 3 receptor 4 subunit) (ITGAX), is a type I transmembrane protein found at high levels on most human dendritic cells, but also on monocytes, macrophages, neutrophils, and some B cells that induces cellular activation and helps trigger neutrophil respiratory burst; expressed in hairy cell leukemias, acute non-lymphocytic leukemias, and some B-cell chronic lymphocytic leukemias.
Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: HC1/1
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human