Tlačiť
Cut resistant gloves, HyFlex® 74-718
Dodávateľ: Ansell
74-718/7 74-718/8 74-718/11 74-718/9 74-718/10
112-4626EA 352.59 EUR
112-4626 112-4627 112-4625 112-4628 112-4624
Cut resistant gloves, HyFlex® 74-718
Rukavice
These knitted gloves are made from stainless steel, spandex, glass fiber and Dyneema® yarn and provide reliable cut protection.
  • Zonal plating along with Dyneema® Diamond Technology enhances protection in high-risk areas of the hand
  • Incorporates Tuff Cuff™ II technology to retain ergonomic shape during use and after laundering
  • KVSD (Knitted Variable Stitch Design) technology to reduce hand fatigue
  • Advanced fibres to help control moisture and hand temperature in cold and warm environments
Ideal for slicing and dicing, filleting fish and food preparation, processing and handling.
Certifikácie: EN 388:2016 (4X4XE); Cat. II
Food contact approved