Tlačiť
HPLC columns, ZORBAX® 300 Å StableBond®
Dodávateľ: AGILENT HEWLETT PACKARD
863973-902 883750-906 883750-909 5065-9911 861973-906
HEWL863973-902EA 1110.35 EUR
HEWL863973-902 HEWL883750-906 HEWL883750-909 HEWL5065-9911 HEWL861973-906
HPLC columns, ZORBAX® 300 Å StableBond®
Kolony chromatografické
These columns are ideal for the reproductible separation of proteins and peptides or other large molecules.
  • Unmatched in their durability at low pH (1 to 8)
  • Temperature use 80 to 90 °C