Tlačiť
Fusion crucibles, graphite
Dodávateľ: Thermo Fisher Scientific
14747.UB 40794.KD
14747.UBEA 567.19 EUR
14747.UB 40794.KD
Fusion crucibles, graphite
Kelímky
These crucibles are manufactured from unpurified extruded graphite.
  • Exceptional thermal uniformity
  • Exactness to dimensional specifications