Tlačiť

Sodium cyclamate 99%

Dodávateľ: Thermo Fisher Scientific
Varování

Synonymá: Sodium N-cyclohexylsulphamate, Sodium N-Cyclohexylsulphamate, N-Cyclohexylsulphamic acid sodium salt

389280010 389280500 389282500
ACRO389280010EA 188.66 EUR
ACRO389280010 ACRO389280500 ACRO389282500
Sodium cyclamate 99%
Sodium cyclamate
Vzorec: C₆H₁₂NNaO₃S
Mw: 201.22
Teplota tavenia: 265
Teplota vzplanutia: >200 °C
Skladovacia teplota: Ambient
Číslo MDL: MFCD00003827
Číslo CAS: 139-05-9
Merck Index: 12,02770

Order Now

Výsledky špecifikačného testu

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤1 %
pH 5.5 to 7.5 (10 % solution)
Titration with HClO4 ≥98.5 % (On dry substance)
Sulfate (SO4) ≤0.1 %
Heavy metals (as Pb) ≤10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR