Tlačiť
ESD safety shoes, lace-up, Fun, Maverick Red Low
Dodávateľ: ELTEN
723341/38 723341/39 723341/40 723341/41 723341/42 723341/43 723341/44 723341/45 723341/46 723341/47 723341/48
114-7668EA 100.68 EUR
114-7668 114-7669 114-7670 114-7671 114-7672 114-7673 114-7674 114-7675 114-7676 114-7677 114-7678
ESD safety shoes, lace-up, Fun, Maverick Red Low
Obuv
Sporty, lightweight leather-free ESD safety shoe made from microfibre and mesh inserts, a padded tongue plus a slip resistant, dual-density PU/PU sole (Fun).
  • Climate-regulating, breathable textile lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), black
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Red/white
Certifikácie: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC.