Tlačiť
ESD safety shoes, lace-up, L10, Vintage Low
Dodávateľ: ELTEN
721071/37 721071/38 721071/39 721071/40 721071/41 721071/42 721071/43 721071/44 721071/45 721071/46 721071/47
114-7636EA 107.1 EUR
114-7636 114-7637 114-7638 114-7639 114-7640 114-7641 114-7642 114-7643 114-7644 114-7645 114-7646
ESD safety shoes, lace-up, L10, Vintage Low
Obuv
Casual, vintage style, environmental friendly manufactured ESD safety shoe made from waxy cowhide leather, a padded tongue plus a low profile, dual-density TPU/PU sole (L10).
  • Bioactive fabric lining
  • Replaceable insole; orthopaedic insoles possible
  • Full length inlay sole (ESD Pro), grey
  • Metal-free, puncture resistant midsole
  • Protective steel toecap (200 J)
Ideal for use in areas where exposure to moisture is expected and where there is a risk of penetration from pointed and sharp objects.
Colour: Grey
Certifikácie: EN 61340-5-1 (ESD); DGUV 112-191; EN ISO 20345; S3, SRC