Ac-DEVD-CHO
Dodávateľ: Biotium
Ac-DEVD-CHO is a synthetic tetrapeptide competitive inhibitor for Caspase-3/7, and contains the amino acid sequence of the PARP cleavage site.
- Soluble in DMSO
- Store at −20 °C
Ac-DEVD-CHO can be used to inhibit Caspase-3/7 activity in apoptotic cells, and to study events downstream of Caspase-3/7 activation.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...