Tlačiť

Sodium-2-bromoethanesulphonate 98%

Dodávateľ: Thermo Fisher Scientific
Varování

Synonymá: Sodium 2-bromoethanesulphonate, 2-Bromoethanesulphonic Acid Sodium Salt

160030250 160031000
ACRO160030250EA 40.86 EUR
ACRO160030250 ACRO160031000
Sodium-2-bromoethanesulphonate 98%
Sodium-2-bromoethanesulphonate

Appearance: White to off-white Crystalline powder

Vzorec: C₂H₅BrO₃S
Mw: 211.00 g/mol
Teplota tavenia: 282…286 °C
Skladovacia teplota: Ambient
Číslo MDL: MFCD00007530
Číslo CAS: 4263-52-9

Order Now

Výsledky špecifikačného testu

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 282 to 286 °C
Titration Argentometric ≥97.5 % (ex Br) On dry substance
Loss on drying ≤0.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Viac About VWR