Tlačiť
Mixing measuring cylinders, tall form, class B, borosilicate glass
Dodávateľ: KAVALIER
632432250718 632432250819 632432250923 632432251125 632432251130 632432251238 632432251343 632432251444 632432251546 632432626018 632432626019 632432626023 632432626030 632432626038 632432626043 632432626044 632432626046
VTRB632432250718EA 0 EUR
VTRB632432250718 VTRB632432250819 VTRB632432250923 VTRB632432251125 VTRB632432251130 VTRB632432251238 VTRB632432251343 VTRB632432251444 VTRB632432251546 VTRB632432626018 VTRB632432626019 VTRB632432626023 VTRB632432626030 VTRB632432626038 VTRB632432626043 VTRB632432626044 VTRB632432626046
Mixing measuring cylinders, tall form, class B, borosilicate glass
Válce Válce odměrné
Borosilicate glass 3.3, class B
  • Hexagonal glass base
  • Brown graduations
  • Avaliable with glass or plastic stopper
Certifikácie: DIN ISO 4788