Tlačiť
Volumetric flasks, class A, blue markings
Dodávateľ: KAVALIER
632431022018 632431022019 632431022023 632431022025 632431022030 632431022037 632431022038 632431022043 632431022044 632431022046
VTRB632431022018EA 0 EUR
VTRB632431022018 VTRB632431022019 VTRB632431022023 VTRB632431022025 VTRB632431022030 VTRB632431022037 VTRB632431022038 VTRB632431022043 VTRB632431022044 VTRB632431022046
Volumetric flasks, class A, blue markings
Baňky Baňky odměrné
Borosilicate glass, clear, with beaded rim.
  • Individually calibrated
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Certifikácie: DIN EN ISO 1042
Informace o dodávce: Supplied with verification certificate.