Tlačiť
Volumetric flask, class A, blue markings
Dodávateľ: GLASSCO
132.202.01 132.202.02 132.202.03 132.202.04 132.202.05 132.202.06 132.202.07 132.202.08 132.202.09 132.202.10 132.202.11
GLAS132.202.01EA 4.65 EUR
GLAS132.202.01 GLAS132.202.02 GLAS132.202.03 GLAS132.202.04 GLAS132.202.05 GLAS132.202.06 GLAS132.202.07 GLAS132.202.08 GLAS132.202.09 GLAS132.202.10 GLAS132.202.11
Volumetric flask, class A, blue markings
Baňky Baňky odměrné
Borosilicate glass 3.3, clear, with beaded rim.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
  • With batch number and LOT certificate
Certifikácie: According to DIN 12664 and ISO 1042.