Tlačiť
Pipette tips, extended length, ART™
Dodávateľ: Thermo Fisher Scientific
3510 3540-11 3151-HR 3152-HR 3511-HR 3512-HR 3541-HR 3542-HR 3791-HR 3792-HR
612-9750EA 19.33 EUR
612-9750 732-2249 613-2455 613-2457 613-2467 613-2469 613-2479 613-2488 613-2782 613-2784
Pipette tips, extended length, ART™
Špičky pipetovací
These non filtered tips are designed to fit virtually any micropipette. They are suitable for use when working with the most difficult samples, such as radioactive compounds, viscous materials, fragile cell lines, viruses, difficult proteins and PCR reagents.
  • Non sterile tips are certified to be free of RNase, DNase, DNA, ATP, endotoxins, pyrogens and PCR inhibitors
  • Sterile tips are certified to be free of RNase, DNase, DNA, ATP, bioburden, endotoxins, pyrogens and PCR inhibitors
  • Extended length to prevent contamination by keeping the pipettete shaft from entering the sample vessel
  • Rack cover can easily be realigned from a traditional lift-off to a hinged rack for one-handed access to tips
  • Front latch closes and locks ensure safe and secure storage
Informace o dodávce: Tips are available in bulk packaging or in racks.