Tlačiť
VWR®, Cover Glasses, Square
Dodávateľ: VWR Collection
631-1566EA 33.21 EUR
631-1566 631-1567 631-1568 631-1570 631-1571
VWR®, Cover Glasses, Square
Sklíčka krycí
These cover slips are specially designed for diagnostic applications.
  • Transparent hydrolytic class 1 glass
Balenie: In dispenser boxes with separate lid.