Tlačiť
Ringer's solution (quarter strength) standard, unbuffered
Dodávateľ: Thermo Fisher Scientific
J67524.AP J67524.K2
J67524.APEA 133.98 EUR
J67524.AP J67524.K2
Ringer's solution (quarter strength) standard, unbuffered
Ringer's solution (quarter strength)
Contains 147 mM sodium chloride, 4 mM potassium chloride, and 3 mM calcium chloride in endotoxin free water and sterile filtered through 0.22 micron filter.