5,7-Dichlorokynurenic acid monohydrate
Dodávateľ: Thermo Fisher Scientific
CAS No.: 190908-40-8
Molecular Formula: C10H5Cl2NO3.H2O
Formula Weight: 276.08 (258.06anhy)
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Pale yellow
Solubility: Soluble in 1 eq. NaOH to 50 mM
MDL No.: MFCD11045951
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...