Boc-HYP-OBZL 95%
Dodávateľ: Thermo Fisher Scientific
Synonymá:
N-Boc-trans-4-hydroxy-L-proline benzyl ester, (2S,4R)-2-Benzyl 1-tert-butyl 4-hydroxypyrrolidine-1,2-dicarboxylate
H62495.03
H62495.06
H62495.03EA
121.37
EUR
H62495.03
H62495.06
Boc-HYP-OBZL 95%
Boc-Hyp-OBZL
Purity: 95%
CAS No.: 89813-47-8
Molecular Formula: C17H23NO5
Formula Weight: 321.37
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Pale brown
MDL No.: MFCD00076979
Vzorec:
C₁₇H₂₃NO₅ Mw: 321.373 g/mol |
Číslo MDL:
MFCD00076979 Číslo CAS: 89813-47-8 |