1-Cyclopentenecarbonitrile 97%
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
H25836.06
H25836.03
H25836.06EA
313.64
EUR
H25836.06
H25836.03
1-Cyclopentenecarbonitrile 97%
1-Cyclopentenecarbonitrile
Purity: 97%
CAS No.: 3047-38-9
Molecular Formula: C6H7N
Formula Weight: 93.14
Refractive Index: 1.4715
Boiling Point: 72°/15mm
UN No.: UN3276
MDL No.: MFCD00043765
Vzorec:
C₆H₇N Mw: 93.14 g/mol Teplota varu: 72 °C (15 mmHg) Hustota: 0.944 Skladovacia teplota: Ambient |
Číslo MDL:
MFCD00043765 Číslo CAS: 3047-38-9 OSN: 3276 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...