2-Cyanothioacetamide 98%
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
A15436.06
A15436.14
A15436.22
A15436.06EA
69.7
EUR
A15436.06
A15436.14
A15436.22
2-Cyanothioacetamide 98%
2-Cyanothioacetamide
Purity: 98%
CAS No.: 7357-70-2
Molecular Formula: NCCH2CSNH2
Formula Weight: 100.14
Melting Point: 114-120°
Beilstein Reference: 1699934
UN No.: UN3439
MDL No.: MFCD00010025
Vzorec:
C₃H₄N₂S Mw: 100.139 g/mol Teplota tavenia: 114…120 °C Skladovacia teplota: Ambient |
Číslo MDL:
MFCD00010025 Číslo CAS: 7357-70-2 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...