Nickel iron molybdenum, foil, annealed, Thickness: 0.102 mm (0.004 in)
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
45092.HB
45092.4K
45092.HBEA
121.54
EUR
45092.HB
45092.4K
Nickel iron molybdenum, foil, annealed, Thickness: 0.102 mm (0.004 in)
Nickel iron molybdenum
Molecular Formula: Ni:Fe:Mo; 80:15:4.2 wt%
Physical Form: ≈40.1g/150mmx300mm
MDL No.: MFCD09910537