Coal tar solution contains 81.0-86.0% alcohol
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
43671.K4
43671.AP
43671.K4EA
225.83
EUR
43671.K4
43671.AP
Coal tar solution contains 81.0-86.0% alcohol
Coal tar
CAS No.: 8007-45-2
Flash Point: 13°(55°F)
Physical Form: Liquid
Appearance: Dark brown
Odour: Alcohol-like
UN No.: UN1993
MDL No.: MFCD00146482
Teplota vzplanutia:
13 °C (55 °F) Skladovacia teplota: Ambient |
Číslo MDL:
MFCD00146482 Číslo CAS: 8007-45-2 OSN: 1993 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...