Ruthenium(III) nitrosylsulphate ≥99.99% (metals basis), Premion®
Dodávateľ: Thermo Fisher Scientific
|
Varování
|
43436.03
43436.06
43436.03EA
168.07
EUR
43436.03
43436.06
Ruthenium(III) nitrosylsulphate ≥99.99% (metals basis), Premion®
Ruthenium(III) nitrosylsulphate
Purity: 99.99%
Molecular Formula: [Ru(NO)]2(SO4)3
Formula Weight: 550.34
Physical Form: Powder
Solubility: Soluble in water
MDL No.: MFCD02684508
Applications: Electroplating, coating