Samarium cobalt (33% Sm), REacton®
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
42732.18
42732.30
42732.18EA
127.38
EUR
42732.18
42732.30
Samarium cobalt (33% Sm), REacton®
Samarium cobalt
CAS No.: 12017-68-4
Molecular Formula: SmCo5, Intermetallic, Magnet Alloy Additive
Physical Form: Fine Powder
Sensitivity: Air and Moisture Sensitive
Odour: Odourless
UN No.: UN3089
MDL No.: MFCD02091682
Vzorec:
CoSm Mw: 209.293 g/mol Skladovacia teplota: Ambient |
Číslo MDL:
MFCD02091682 Číslo CAS: 12017-68-4 EINECS: 234-625-7 OSN: 3089 ADR: 4.1 III |