Chromyl chloride ≥99.99% (metals basis)
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
40517.09
40517.18
40517.09EA
147.61
EUR
40517.09
40517.18
Chromyl chloride ≥99.99% (metals basis)
Chromyl chloride
Purity: 99.99%
CAS No.: 14977-61-8
Molecular Formula: CrO2Cl2
Formula Weight: 154.90
Physical Form: Liquid
Density: 1.911
Sensitivity: Moisture Sensitive
Odour: Pungent, makes eyes water
Melting Point: -96.5°
Boiling Point: 117°
UN No.: UN1758
MDL No.: MFCD00010951
Applications: Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins
Vzorec:
CrO₂Cl₂ Teplota varu: 117 °C Teplota tavenia: –96,5 °C Hustota: 1,911 Skladovacia teplota: Ambient |
Číslo MDL:
MFCD00010951 Číslo CAS: 14977-61-8 EINECS: 239-056-8 OSN: 1758 ADR: 8,I Merck Index: 14,2248 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...