Tlačiť
Crystal polishing kit
Dodávateľ: Thermo Fisher Scientific
39798.KT 39806.LA
39798.KTEA 841.16 EUR
39798.KT 39806.LA
Crystal polishing kit
Zařízení pro leštění a tromlování
The Crystal Polishing Kit contains everything needed by the spectroscopist for repolishing and buffing most infrared transmission crystals, including KRS-5. Two ground glass discs are provided – one for fine grinding and the other to which self-stick polishing pads may be affixed.
Kit contents:
Silicon carbide, 400 and 600 grinding compound
Cerium oxide polishing compound
Aluminium oxide polishing compound (ideal for resurfacing KRS-5 crystals)
Finger cots
3× Camel hair brushes
2× Polishing pads
2× Ground glass plates
5× Polyethylene squeeze bottles with the above-mentioned compounds contained therein
Fitted mahogany base, and instructions