Lithium tartrate monohydrate ≥99%
Dodávateľ: Thermo Fisher Scientific
39357.18
39357.30
39357.18EA
89.05
EUR
39357.18
39357.30
Lithium tartrate monohydrate ≥99%
Lithium tartrate monohydrate
Purity: 99%
CAS No.: 6108-32-3
Molecular Formula: C4H4Li2O6.H2O
Formula Weight: 179.97 (161.97anhy)
Physical Form: Powder/Aggregates
Odour: Odourless
Solubility: Soluble in water
MDL No.: MFCD00054421
Vzorec:
C₄H₄Li₂O₆·H₂O Skladovacia teplota: Ambient |
Číslo MDL:
MFCD00054421 Číslo CAS: 6108-32-3 EINECS: 212-772-8 |