Tlačiť
Platina 3% on activated carbon wetted with ca. 50% water, powder, reduced
Dodávateľ: Thermo Fisher Scientific
Synonymá: Platinium
38316.14 38316.06
38316.14EA 364.51 EUR
38316.14 38316.06
Platina 3% on activated carbon wetted with ca. 50% water, powder, reduced
Platina
CAS No.: 7440-06-4 Physical Form: 900-1100 m²/g MDL No.: MFCD00011179
Objednávacie informácie: Sold on a dry weight basis. Unit weight excludes water weight
Vzorec: Pt
Mw: 195.08
Teplota varu: 3827
Teplota tavenia: 1769
Skladovacia teplota: Ambient
Číslo MDL: MFCD00011179
Číslo CAS: 7440-06-4
Merck Index: 12,07689