Gold nickel ≥99.85% (metals basis), wire, annealed, Ø 0.5 mm (0.02 in)
Dodávateľ: Thermo Fisher Scientific
|
Varování
|
36451.BS
36451.BW
36451.G1
36451.BSEA
206.58
EUR
36451.BS
36451.BW
36451.G1
Gold nickel ≥99.85% (metals basis), wire, annealed, Ø 0.5 mm (0.02 in)
Gold nickel
Purity: 99.85%
Molecular Formula: Au:Ni; 82:18 wt%
Physical Form: ≈0.32g/10cm, Temper: soft
MDL No.: MFCD02091707
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...