Potassium bromate/Potassium bromide 0.1 N standardised solution
Dodávateľ: Thermo Fisher Scientific
|
Nebezpečí
|
35593.K2
35593.K7
35593.K2EA
116.73
EUR
35593.K2
35593.K7
Potassium bromate/Potassium bromide 0.1 N standardised solution
Potassium bromate/Potassium bromide
Molecular Formula: KBrO3/KBr
Physical Form: Liquid
MDL No.: MFCD00011359