Tlačiť
Dissolved oxygen bottles
Dodávateľ: VITRUM GLASS
2772/A100-150 2772/A250-300
VTRA2772/A100-150EA 25.24 EUR
VTRA2772/A100-150 VTRA2772/A250-300
Dissolved oxygen bottles
Láhve Lahvičky na stanovení rozpuštěného kyslíku
Glass, clear.
  • With white labelling area
  • Oblique stopper
  • Marked with accurate volume
For the determination of dissolved oxygen in water according to Winkler.