Tlačiť
Irrigators, barrel shaped, tubulated
Dodávateľ: KAVALIER
632415623952 632415623956 632415623966 632415623968
VTRB632415623952EA 0 EUR
VTRB632415623952 VTRB632415623956 VTRB632415623966 VTRB632415623968
Irrigators, barrel shaped, tubulated
Láhve Aspirátory
Borosilicate glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With loose drop-on cover with knob. These bottles are provided with standardised ground neck near bottom and bleeding cock.
Informace o dodávce: Each bottle comes with lid, tap and tube at the bottom.