Tlačiť
Dropping bottles, glass, with ground-in pipette stopper
Dodávateľ: VITRUM GLASS
2016/B/50 2016/B/100 2016/B/250 2016/H/50 2016/H/100 2016/H/250
VTRA2016/B/50EA 33.45 EUR
VTRA2016/B/50 VTRA2016/B/100 VTRA2016/B/250 VTRA2016/H/50 VTRA2016/H/100 VTRA2016/H/250
Dropping bottles, glass, with ground-in pipette stopper
Láhve Lahvičky kapací
Soda-lime glass, clear or amber.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Replaceable NS glass pipette with rubber suction bulb
Amber to protect light-sensitive substances.