Tlačiť
Distillation flasks, with side arm
Dodávateľ: KAVALIER
632425220025 632425220050 632425220500 632425220940
VTRB632425220025EA 16.02 EUR
VTRB632425220025 VTRB632425220050 VTRB632425220500 VTRB632425220940
Distillation flasks, with side arm
Baňky Baňky destilační
Borosilicate glass 3.3, SIMAX®.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Flasks with side arm sealed at the bottom third part of neck. Uniform wall thickness distribution makes these flasks ideal for heating applications and distillations.